siRNA Details
| Gene Symbol: BRD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009877.2 | NP_001009877.2 | 6 | 699 | ||
| NM_001317951.1 | NP_001304880.1 | 6 | 991 | |||
| NM_023924.4 | NP_076413.3 | 6 | 872 | |||
| NR_134296.1 | 6 | 1208 | ||||
| XM_006714494.3 | XP_006714557.1 | 6 | 851 | |||
| XM_017009780.1 | XP_016865269.1 | 6 | 851 | |||
| GenBank mRNA | AK023503.1 | - | 6 | 969 | ||
| AK024392.1 | - | 3 | 707 | |||
| AK297573.1 | - | 6 | 686 | |||
| AK299157.1 | - | 6 | 872 | |||
| AK301987.1 | - | 6 | 823 | |||
| BC007217.1 | - | 6 | 888 | |||
| BC041590.1 | - | 4 | 558 | 5428011 | ||
| BX648703.1 | - | 6 | 1210 | |||
| DQ248311.1 | - | 2 | 197 | |||

BRD9
Genomic Map