siRNA Details
| Gene Symbol: MRPL57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024026.4 | NP_076931.1 | 2 | 1367 | ||
| GenBank mRNA | BC023616.2 | - | 2 | 924 | 4645641 | |
| BC068492.1 | - | 2 | 934 | 4801279 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRPL57| Gene Symbol: MRPL57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024026.4 | NP_076931.1 | 2 | 1367 | ||
| GenBank mRNA | BC023616.2 | - | 2 | 924 | 4645641 | |
| BC068492.1 | - | 2 | 934 | 4801279 | ||