siRNA Details
| Gene Symbol: GDAP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256737.1 | NP_001243666.1 | 6 | 1528 | ||
| NM_001256738.1 | NP_001243667.1 | 6 | 1318 | |||
| NM_001256739.1 | NP_001243668.1 | 5 | 1297 | |||
| NM_001256740.1 | NP_001243669.1 | 4 | 1258 | |||
| NM_024034.4 | NP_076939.3 | 6 | 1471 | |||
| NR_046353.1 | 4 | 1104 | ||||
| GenBank mRNA | AK056251.1 | - | 6 | 1461 | ||
| AK293484.1 | - | 5 | 1247 | |||
| CR749676.1 | - | 2 | 759 | |||

GDAP1L1
Genomic Map