siRNA Details
| Gene Symbol: C19orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024038.2 | NP_076943.1 | 2, 3 | 489 | ||
| GenBank mRNA | AF061732.1 | - | 1, 2 | 65 | ||
| AK027588.1 | - | 2, 3 | 384 | |||
| AK315754.1 | - | 2, 3 | 455 | |||
| BC000216.2 | - | 2, 3 | 489 | 2961319 | ||
| BC004533.2 | - | 2, 3 | 434 | 3139656 | ||
| BC015347.1 | - | 2, 3 | 448 | 4156774 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026760, AK003299, BC023284, AK009653, BC029622.

C19orf43
Genomic Map