siRNA Details
| Gene Symbol: VPS33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022916.4 | NP_075067.2 | 13 | 2570 | ||
| GenBank mRNA | AF439857.1 | - | 13 | 2481 | ||
| AK026048.1 | - | 7 | 1693 | |||
| AK026840.1 | - | 13 | 2480 | |||
| BC016617.1 | - | 13 | 2478 | 3855313 | ||

VPS33A
Genomic Map