siRNA Details
| Gene Symbol: ZNF649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023074.3 | NP_075562.2 | 5 | 2779 | ||
| GenBank mRNA | AL713677.1 | - | 5 | 2721 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF649| Gene Symbol: ZNF649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023074.3 | NP_075562.2 | 5 | 2779 | ||
| GenBank mRNA | AL713677.1 | - | 5 | 2721 | ||