siRNA Details
| Gene Symbol: YEATS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300950.1 | NP_001287879.1 | 5 | 891 | ||
| NM_006530.3 | NP_006521.1 | 7 | 1053 | |||
| GenBank mRNA | AJ245746.1 | - | 7 | 1001 | ||
| AK001413.1 | - | 7 | 826 | |||
| BC000994.2 | - | 7 | 914 | 2899965 | ||
| U61384.1 | - | 7 | 1011 | |||

YEATS4
Genomic Map