siRNA Details
| Gene Symbol: VKORC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001311311.1 | NP_001298240.1 | 4 | 1102 | ||
| NM_024006.5 | NP_076869.1 | 3 | 1018 | |||
| NM_206824.2 | NP_996560.1 | 2 | 908 | |||
| GenBank mRNA | AF176924.1 | - | 4 | 1042 | ||
| AK129513.1 | - | 2 | 706 | |||
| AY466113.1 | - | 3 | 960 | |||
| BC000828.1 | - | 3 | 748 | |||
| BC002911.2 | - | 3 | 786 | 3944530 | ||
| CD249837.1 | - | 710 | ||||

VKORC1
Genomic Map