siRNA Details
| Gene Symbol: YIPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321439.1 | NP_001308368.1 | 10 | 1547 | ||
| NM_001321440.1 | NP_001308369.1 | 9 | 1430 | |||
| NM_024029.4 | NP_076934.1 | 10 | 1818 | |||
| XM_011528270.2 | XP_011526572.1 | 10 | 1564 | |||
| XM_011528271.2 | XP_011526573.1 | 9 | 1408 | |||
| GenBank mRNA | BC013014.2 | - | 10 | 1487 | 2960233 | |
| BX327495.2 | - | 3 | 685 | |||

YIPF2
Genomic Map