siRNA Details
| Gene Symbol: LBHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024099.3 | NP_077004.2 | 4, 5 | 761 | ||
| GenBank mRNA | AK055736.1 | - | 2, 3 | 2352 | ||
| BC001434.2 | - | 4, 5 | 761 | 3138904 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LBHD1| Gene Symbol: LBHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024099.3 | NP_077004.2 | 4, 5 | 761 | ||
| GenBank mRNA | AK055736.1 | - | 2, 3 | 2352 | ||
| BC001434.2 | - | 4, 5 | 761 | 3138904 | ||