siRNA Details
| Gene Symbol: C1orf116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083924.1 | NP_001077393.1 | 3 | 3536 | ||
| NM_023938.5 | NP_076427.2 | 4 | 3722 | |||
| XM_005273259.1 | XP_005273316.1 | 2 | 3290 | |||
| XM_006711530.1 | XP_006711593.1 | 4 | 3599 | |||
| XM_011509973.2 | XP_011508275.1 | 3 | 3392 | |||
| GenBank mRNA | AK096741.1 | - | 1 | 1338 | ||
| BC001943.1 | - | 1 | 220 | |||
| BC002325.2 | - | 1 | 220 | |||

C1orf116
Genomic Map