siRNA Details
| Gene Symbol: RBM42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319113.1 | NP_001306042.1 | 1 | 251 | ||
| NM_024321.4 | NP_077297.2 | 1 | 251 | |||
| XM_005259257.2 | XP_005259314.1 | 1 | 246 | |||
| GenBank mRNA | AK301669.1 | - | 1 | 176 | ||
| BC002868.2 | - | 1 | 190 | 3941786 | ||
| BC004204.2 | - | 1 | 189 | 3532802 | ||
| BC031682.1 | - | 1 | 218 | 5163682 | ||
| HY031737.1 | - | 1 | 252 | |||

RBM42
Genomic Map