siRNA Details
| Gene Symbol: FSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024333.2 | NP_077309.1 | 3 | 338 | ||
| XM_005259644.2 | XP_005259701.1 | 3 | 337 | |||
| GenBank mRNA | AF316829.1 | - | 3 | 229 | ||
| AK021750.1 | - | 3 | 238 | |||
| AK295523.1 | - | 3 | 233 | |||
| AK303606.1 | - | 3 | 222 | |||
| AK315661.1 | - | 3 | 332 | |||
| AY032617.1 | - | 3 | 261 | |||
| BC003124.1 | - | 3 | 238 | 3502614 | ||

FSD1
Genomic Map