siRNA Details
| Gene Symbol: ODF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024410.3 | NP_077721.2 | 1 | 184 | ||
| GenBank mRNA | AK315276.1 | - | 1 | 177 | ||
| BC104456.1 | - | 1 | 155 | 40030623 | ||
| BC104457.1 | - | 1 | 155 | 40030627 | ||
| DB457574.1 | - | 1 | 184 | |||

ODF1
Genomic Map