siRNA Details
| Gene Symbol: PVRIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024070.3 | NP_076975.2 | 6 | 1495 | ||
| XM_011516575.2 | XP_011514877.1 | 7 | 1511 | |||
| GenBank mRNA | BC001129.2 | - | 5 | 1183 | ||
| BC032113.2 | - | 3 | 882 | |||
| BC073861.1 | - | 5 | 1158 | |||
| BC107873.1 | - | 7 | 1520 | 6452536 | ||

PVRIG
Genomic Map