siRNA Details
| Gene Symbol: AEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129.4 | NP_001120.3 | 2 | 593 | ||
| XM_011515162.1 | XP_011513464.1 | 2 | 634 | |||
| GenBank mRNA | AB209744.1 | - | 2 | 547 | ||
| AF053944.1 | - | 2 | 427 | |||
| BC038588.1 | - | 2 | 593 | 5764769 | ||

AEBP1
Genomic Map