siRNA Details
| Gene Symbol: C1orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024097.3 | NP_077002.2 | 2, 3 | 270 | ||
| GenBank mRNA | BC001508.2 | - | 2, 3 | 246 | 2959109 | |
| BC001711.1 | - | 2, 3 | 246 | 3543543 | ||
| CN256666.1 | - | 1, 2 | 54 | |||
| DA684965.1 | - | 2, 3 | 270 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001012400, AK167445, BC035522, BC089614, AK158349.

C1orf50
Genomic Map