siRNA Details
| Gene Symbol: CHAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024111.3 | NP_077016.2 | 3 | 526 | ||
| XM_017022563.1 | XP_016878052.1 | 4 | 5050 | |||
| XM_017022564.1 | XP_016878053.1 | 5 | 2832 | |||
| XM_017022565.1 | XP_016878054.1 | 4 | 5039 | |||
| GenBank mRNA | BC001683.1 | - | 3 | 480 | 2964714 | |
| BC001847.2 | - | 3 | 471 | 2964714 | ||
| BC019625.1 | - | 3 | 482 | 4473135 | ||
| BQ900544.1 | - | 3 | 526 | |||

CHAC1
Genomic Map