siRNA Details
| Gene Symbol: KLF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206.2 | NP_001197.1 | 1 | 1467 | ||
| GenBank mRNA | AB451404.1 | - | 1 | 173 | ||
| AK313631.1 | - | 1 | 248 | |||
| BC069431.1 | - | 1 | 246 | 7262239 | ||
| BC074879.2 | - | 1 | 222 | 30915238 | ||
| BC074880.2 | - | 1 | 222 | 30915472 | ||
| D31716.1 | - | 1 | 1437 | |||

KLF9
Genomic Map