siRNA Details
| Gene Symbol: DSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004949.4 | NP_004940.1 | 3, 4 | 823 | ||
| NM_024422.4 | NP_077740.1 | 3, 4 | 823 | |||
| XM_005258206.4 | XP_005258263.1 | 3, 4 | 324 | |||
| GenBank mRNA | AK290313.1 | - | 3, 4 | 514 | ||
| BC063291.1 | - | 3, 4 | 805 | 30337780 | ||
| BX648425.1 | - | 3, 4 | 375 | |||
| CR749226.1 | - | 3, 4 | 541 | |||
| EF017811.1 | - | 3, 4 | 362 | |||
| EF017812.1 | - | 3, 4 | 362 | |||
| X56807.1 | - | 3, 4 | 402 | |||

DSC2
Genomic Map