siRNA Details
| Gene Symbol: NUP37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024057.3 | NP_076962.2 | 7 | 752 | ||
| GenBank mRNA | AF514994.1 | - | 6 | 606 | ||
| AK026271.1 | - | 6 | 587 | |||
| AK310619.1 | - | 7 | 712 | |||
| BC000861.2 | - | 6 | 670 | 3459639 | ||

NUP37
Genomic Map