siRNA Details
Gene Symbol: HSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040667.2 | NP_001035757.1 | 6, 7 | 1455 | ||
NM_001538.3 | NP_001529.2 | 6, 7 | 1455 | |||
GenBank mRNA | AB029348.1 | - | 4, 5 | 497 | ||
AK225152.1 | - | 4, 5 | 512 | |||
AK296210.1 | - | 4, 5 | 512 | |||
BC130383.1 | - | 4, 5 | 558 | |||
D87673.1 | - | 4, 5 | 497 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011939, NM_001256044, NM_001256042, AK216507, AF160966, and others.
Panther Classification:
Gene Ontology Categories:
