siRNA Details
| Gene Symbol: LENG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024316.2 | NP_077292.2 | 2 | 318 | ||
| GenBank mRNA | AF211966.1 | - | 1 | 96 | ||
| BC014986.1 | - | 2 | 283 | 4877885 | ||
| BP870984.1 | - | 2 | 319 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LENG1| Gene Symbol: LENG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024316.2 | NP_077292.2 | 2 | 318 | ||
| GenBank mRNA | AF211966.1 | - | 1 | 96 | ||
| BC014986.1 | - | 2 | 283 | 4877885 | ||
| BP870984.1 | - | 2 | 319 | |||