siRNA Details
| Gene Symbol: FSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024333.2 | NP_077309.1 | 7, 8 | 861 | ||
| XM_005259644.2 | XP_005259701.1 | 7, 8 | 860 | |||
| GenBank mRNA | AF316829.1 | - | 7, 8 | 752 | ||
| AK021750.1 | - | 7, 8 | 761 | |||
| AK315661.1 | - | 7, 8 | 855 | |||
| AY032617.1 | - | 7, 8 | 784 | |||
| BC003124.1 | - | 7, 8 | 761 | 3502614 | ||
| BC016442.1 | - | 1, 2 | 465 | |||

FSD1
Genomic Map