siRNA Details
| Gene Symbol: TMEM43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024334.2 | NP_077310.1 | 12 | 3210 | ||
| XM_017007176.1 | XP_016862665.1 | 13 | 3332 | |||
| GenBank mRNA | AK027827.1 | - | 7 | 2486 | ||
| AK074073.1 | - | 10 | 5641 | |||
| AL136916.1 | - | 12 | 3103 | |||
| AY358625.1 | - | 13 | 1864 | |||
| BC003125.1 | - | 12 | 3105 | 3503636 | ||

TMEM43
Genomic Map