siRNA Details
| Gene Symbol: ZNF557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044387.1 | NP_001037852.1 | 8 | 1348 | ||
| NM_001044388.1 | NP_001037853.1 | 8 | 1339 | |||
| NM_024341.2 | NP_077317.2 | 8 | 1360 | |||
| GenBank mRNA | AK095524.1 | - | 8 | 1093 | ||
| AK297283.1 | - | 8 | 1117 | |||
| AK315405.1 | - | 8 | 1377 | |||
| BC003020.1 | - | 8 | 1110 | 2822966 | ||
| BC031208.1 | - | 8 | 1117 | |||
| BC058040.1 | - | 8 | 1089 | |||

ZNF557
Genomic Map