siRNA Details
| Gene Symbol: HAUS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303143.1 | NP_001290072.1 | 3 | 1041 | ||
| NM_024511.6 | NP_078787.2 | 2 | 770 | |||
| GenBank mRNA | AF040964.1 | - | 1 | 514 | ||
| AK056134.1 | - | 2 | 1258 | |||
| AK293948.1 | - | 2 | 1258 | |||
| BC003648.2 | - | 3 | 1042 | 3534250 | ||
| BC025356.1 | - | 1 | 282 | 4906369 | ||
| CR749640.1 | - | 2 | 739 | |||

HAUS3
Genomic Map