siRNA Details
| Gene Symbol: NKAIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024522.2 | NP_078798.2 | 4 | 732 | ||
| XM_017002320.1 | XP_016857809.1 | 3 | 326 | |||
| GenBank mRNA | AI884789.1 | - | 4 | 448 | ||
| AK022712.1 | - | 3 | 304 | |||
| AK298874.1 | - | 2 | 223 | |||
| AK299507.1 | - | 4 | 404 | |||
| BC096707.1 | - | 3 | 290 | 40022279 | ||
| BC098173.1 | - | 3 | 290 | 40022282 | ||
| BC098249.1 | - | 3 | 290 | 40022276 | ||
| BC131518.1 | - | 3 | 290 | 40022275 | ||

NKAIN1
Genomic Map