siRNA Details
| Gene Symbol: CARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024537.3 | NP_078813.1 | 3 | 400 | ||
| XM_006719953.2 | XP_006720016.1 | 3 | 480 | |||
| XM_011521116.1 | XP_011519418.1 | 3 | 192 | |||
| XM_011521117.2 | XP_011519419.1 | 3 | 336 | |||
| XM_011521118.2 | XP_011519420.1 | 3 | 336 | |||
| XM_017020741.1 | XP_016876230.1 | 2 | 128 | |||
| XM_017020742.1 | XP_016876231.1 | 3 | 336 | |||
| GenBank mRNA | AB062436.1 | - | 3 | 311 | ||
| AK022180.1 | - | 3 | 336 | |||
| AK301907.1 | - | 2 | 505 | |||
| BC007220.1 | - | 3 | 323 | 2822557 | ||
| DA966145.1 | - | 3 | 355 | |||
| HY150557.1 | - | 3 | 401 | |||
| HY160154.1 | - | 3 | 382 | |||

CARS2
Genomic Map