siRNA Details
| Gene Symbol: RNF128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024539.3 | NP_078815.3 | 2 | 651 | ||
| NM_194463.1 | NP_919445.1 | 2 | 814 | |||
| GenBank mRNA | AF394689.1 | - | 2 | 574 | ||
| AK074264.1 | - | 2 | 814 | |||
| AK126553.1 | - | 2 | 634 | |||
| BC036901.1 | - | 2 | 662 | |||
| BC063404.1 | - | 2 | 653 | 5191785 | ||

RNF128
Genomic Map