siRNA Details
| Gene Symbol: SZT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015284.3 | NP_056099.3 | 61 | 8651 | ||
| XM_005270686.3 | XP_005270743.1 | 63 | 8883 | |||
| XM_006710501.3 | XP_006710564.1 | 62 | 8770 | |||
| XM_011541106.2 | XP_011539408.1 | 62 | 8921 | |||
| XM_011541107.2 | XP_011539409.1 | 59 | 8418 | |||
| XM_017000819.1 | XP_016856308.1 | 63 | 8730 | |||
| XM_017000820.1 | XP_016856309.1 | 62 | 8712 | |||
| XM_017000821.1 | XP_016856310.1 | 59 | 8404 | |||
| GenBank mRNA | AB007936.2 | - | 47 | 6503 | ||
| AK027078.1 | - | 2 | 196 | |||
| BC041069.1 | - | 3 | 332 | |||
| BC052802.1 | - | 4 | 346 | |||
| BC082968.1 | - | 4 | 351 | |||
| BC151232.1 | - | 47 | 6503 | 8860124 | ||

SZT2
Genomic Map