siRNA Details
| Gene Symbol: CHMP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024591.4 | NP_078867.2 | 2 | 178 | ||
| XM_005257668.1 | XP_005257725.1 | 2 | 178 | |||
| GenBank mRNA | AK021811.1 | - | 2 | 116 | ||
| AK223187.1 | - | 2 | 140 | |||
| AK292105.1 | - | 2 | 144 | |||
| AY329087.1 | - | 2 | 100 | |||
| BC010108.2 | - | 2 | 124 | 3604039 | ||
| BP196406.1 | - | 2 | 178 | |||
| CR457284.1 | - | 2 | 100 | |||

CHMP6
Genomic Map