siRNA Details
| Gene Symbol: EFCAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024593.3 | NP_078869.1 | 2 | 295 | ||
| XM_005251303.1 | XP_005251360.1 | 2 | 252 | |||
| XM_011517589.1 | XP_011515891.1 | 2 | 252 | |||
| XM_011517590.2 | XP_011515892.1 | 3 | 425 | |||
| XM_011517591.2 | XP_011515893.1 | 3 | 398 | |||
| XM_017013827.1 | XP_016869316.1 | 4 | 548 | |||
| XM_017013828.1 | XP_016869317.1 | 2 | 827 | |||
| GenBank mRNA | AK021829.1 | - | 2 | 153 | ||
| BC025676.1 | - | 2 | 216 | 5203436 | ||

EFCAB1
Genomic Map