siRNA Details
| Gene Symbol: RPAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146075.1 | NP_001139547.1 | 2 | 238 | ||
| NM_024604.2 | NP_078880.2 | 2 | 238 | |||
| GenBank mRNA | AK025561.1 | - | 2 | 168 | ||
| BC003680.1 | - | 2 | 211 | |||
| BC056415.1 | - | 2 | 163 | 5218249 | ||
| DA096405.1 | - | 2 | 238 | |||

RPAP3
Genomic Map