siRNA Details
| Gene Symbol: PLEKHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024613.3 | NP_078889.1 | 2 | 450 | ||
| GenBank mRNA | AF434819.1 | - | 1 | 126 | ||
| AK023249.1 | - | 2 | 223 | |||
| AL834473.1 | - | 2 | 373 | |||
| BC011806.2 | - | 2 | 350 | 4335031 | ||

PLEKHF2
Genomic Map