siRNA Details
| Gene Symbol: GLB1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286423.1 | NP_001273352.1 | 12 | 1347 | ||
| NM_001286427.1 | NP_001273356.1 | 9 | 864 | |||
| NM_024506.4 | NP_078782.3 | 12 | 1335 | |||
| XM_011511813.2 | XP_011510115.1 | 10 | 924 | |||
| XM_017004895.1 | XP_016860384.1 | 10 | 1031 | |||
| GenBank mRNA | AK055098.1 | - | 7 | 522 | ||
| AK074393.1 | - | 6 | 515 | |||
| AK299404.1 | - | 11 | 1096 | |||
| AK300215.1 | - | 9 | 847 | |||
| AY358639.1 | - | 12 | 1315 | |||

GLB1L
Genomic Map