siRNA Details
| Gene Symbol: RNF219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024546.3 | NP_078822.3 | 4 | 472 | ||
| XM_011535225.1 | XP_011533527.1 | 3 | 322 | |||
| XM_017020743.1 | XP_016876232.1 | 4 | 2267 | |||
| GenBank mRNA | AK098640.1 | - | 4 | 484 | ||
| AL136651.1 | - | 4 | 444 | |||
| BC136764.1 | - | 4 | 467 | 9020754 | ||
| BC136765.1 | - | 4 | 467 | 9020755 | ||
| BI550796.1 | - | 4 | 390 | |||
| CR936752.1 | - | 4 | 449 | |||
| CR936754.1 | - | 4 | 459 | |||
| CR936764.1 | - | 4 | 446 | |||

RNF219
Genomic Map