siRNA Details
| Gene Symbol: TCTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024549.5 | NP_078825.2 | 6, 7 | 944 | ||
| XM_011538733.2 | XP_011537035.1 | 6, 7 | 848 | |||
| XM_017019969.1 | XP_016875458.1 | 6, 7 | 813 | |||
| GenBank mRNA | AK055891.1 | - | 6, 7 | 858 | ||
| BC040113.1 | - | 6, 7 | 834 | 6054789 | ||

TCTN1
Genomic Map