siRNA Details
| Gene Symbol: TMEM53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300746.1 | NP_001287675.1 | 2 | 1231 | ||
| NM_001300747.1 | NP_001287676.1 | 3 | 1263 | |||
| NM_001300748.1 | NP_001287677.1 | 3 | 1260 | |||
| NM_024587.3 | NP_078863.2 | 3 | 1353 | |||
| XM_011542138.2 | XP_011540440.1 | 3 | 1482 | |||
| GenBank mRNA | AK026006.1 | - | 3 | 1160 | ||
| AK225405.1 | - | 3 | 1160 | |||
| AK295356.1 | - | 2 | 953 | |||
| AK310016.1 | - | 3 | 1297 | |||
| AL832539.1 | - | 2 | 1065 | |||
| BC064520.1 | - | 3 | 1172 | 5582377 | ||

TMEM53
Genomic Map