siRNA Details
| Gene Symbol: PANK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024594.3 | NP_078870.1 | 2 | 572 | ||
| GenBank mRNA | AK022961.1 | - | 2 | 317 | ||
| AK223285.1 | - | 2 | 301 | |||
| BC013705.1 | - | 2 | 546 | 4343270 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PANK3| Gene Symbol: PANK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024594.3 | NP_078870.1 | 2 | 572 | ||
| GenBank mRNA | AK022961.1 | - | 2 | 317 | ||
| AK223285.1 | - | 2 | 301 | |||
| BC013705.1 | - | 2 | 546 | 4343270 | ||