siRNA Details
| Gene Symbol: NKAIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024522.2 | NP_078798.2 | 7 | 2546 | ||
| XM_017002320.1 | XP_016857809.1 | 6 | 2140 | |||
| GenBank mRNA | AK022712.1 | - | 6 | 2118 | ||
| AL832547.1 | - | 4 | 1811 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NKAIN1| Gene Symbol: NKAIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024522.2 | NP_078798.2 | 7 | 2546 | ||
| XM_017002320.1 | XP_016857809.1 | 6 | 2140 | |||
| GenBank mRNA | AK022712.1 | - | 6 | 2118 | ||
| AL832547.1 | - | 4 | 1811 | |||