siRNA Details
| Gene Symbol: RNF128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024539.3 | NP_078815.3 | 7 | 2391 | ||
| NM_194463.1 | NP_919445.1 | 7 | 2554 | |||
| GenBank mRNA | AK027169.1 | - | 5 | 1563 | ||
| AK074264.1 | - | 7 | 2554 | |||
| AK126553.1 | - | 7 | 2374 | |||
| BC030951.1 | - | 6 | 1667 | |||
| BC036901.1 | - | 7 | 2403 | |||
| BC056677.1 | - | 5 | 1538 | |||
| BC063404.1 | - | 7 | 2393 | 5191785 | ||

RNF128
Genomic Map