siRNA Details
| Gene Symbol: C10orf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024541.2 | NP_078817.2 | 26 | 3450 | ||
| XM_006717969.1 | XP_006718032.1 | 24 | 3238 | |||
| XM_011540151.2 | XP_011538453.1 | 26 | 3495 | |||
| XM_011540152.2 | XP_011538454.1 | 25 | 3382 | |||
| XM_011540153.2 | XP_011538455.1 | 25 | 3346 | |||
| XM_011540154.2 | XP_011538456.1 | 24 | 3280 | |||
| XM_011540156.2 | XP_011538458.1 | 19 | 3414 | |||
| XM_011540157.2 | XP_011538459.1 | 25 | 3399 | |||
| GenBank mRNA | AK023176.1 | - | 9 | 2079 | ||
| AK097972.1 | - | 3 | 1813 | |||

C10orf76
Genomic Map