siRNA Details
| Gene Symbol: RNF219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024546.3 | NP_078822.3 | 6 | 1111 | ||
| XM_011535225.1 | XP_011533527.1 | 5 | 961 | |||
| XM_017020743.1 | XP_016876232.1 | 6 | 2906 | |||
| GenBank mRNA | AK023511.1 | - | 1 | 300 | ||
| AK098640.1 | - | 7 | 1199 | |||
| AL136651.1 | - | 6 | 1083 | |||
| BC136764.1 | - | 6 | 1106 | 9020754 | ||
| BC136765.1 | - | 6 | 1106 | 9020755 | ||
| CR936752.1 | - | 6 | 1088 | |||
| CR936754.1 | - | 6 | 1099 | |||
| CR936764.1 | - | 7 | 1015 | |||

RNF219
Genomic Map