siRNA Details
| Gene Symbol: ACSS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024560.2 | NP_078836.1 | 14 | 1866 | ||
| XM_005269150.2 | XP_005269207.1 | 14 | 1800 | |||
| XM_011538740.2 | XP_011537042.1 | 14 | 1515 | |||
| GenBank mRNA | AK025616.1 | - | 14 | 1817 | ||
| AK074267.1 | - | 4 | 290 | |||
| AK074938.1 | - | 9 | 1047 | |||
| AX764480.1 | - | 14 | 2016 | |||
| AX764482.1 | - | 14 | 1772 | |||
| BC009317.2 | - | 14 | 1866 | 4120563 | ||
| BC015769.1 | - | 14 | 1866 | 4902942 | ||

ACSS3
Genomic Map