siRNA Details
Gene Symbol: GAL3ST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024637.4 | NP_078913.3 | 3, 4 | 814 | ||
GenBank mRNA | AF289575.1 | - | 2, 3 | 609 | ||
AF316113.1 | - | 3, 4 | 657 | |||
AK022178.1 | - | 2, 3 | 453 | |||
AK092367.1 | - | 2, 3 | 1440 | |||
AK225890.1 | - | 2, 3 | 590 | |||
AL833824.1 | - | 2, 3 | 459 | |||
BC012976.2 | - | 3, 4 | 633 | 3636329 |
