siRNA Details
| Gene Symbol: TBL1XR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321193.1 | NP_001308122.1 | 4 | 440 | ||
| NM_001321194.1 | NP_001308123.1 | 5 | 620 | |||
| NM_001321195.1 | NP_001308124.1 | 3 | 337 | |||
| NM_024665.5 | NP_078941.2 | 4 | 501 | |||
| XM_005247775.2 | XP_005247832.1 | 3 | 456 | |||
| XM_006713746.1 | XP_006713809.1 | 5 | 510 | |||
| XM_011513141.1 | XP_011511443.1 | 5 | 444 | |||
| XM_011513142.2 | XP_011511444.1 | 4 | 425 | |||
| XM_011513143.2 | XP_011511445.1 | 3 | 351 | |||
| XM_017007185.1 | XP_016862674.1 | 4 | 438 | |||
| GenBank mRNA | AF268193.1 | - | 4 | 334 | ||
| AF314544.1 | - | 3 | 300 | |||
| AK296946.1 | - | 3 | 235 | |||
| BC060320.1 | - | 3 | 354 | |||
| BC113421.1 | - | 3 | 243 | 8322473 | ||
| BX648935.1 | - | 4 | 435 | |||
| DA159105.1 | - | 3 | 286 | |||
| DA460721.1 | - | 2 | 262 | |||
| DB150466.1 | - | 5 | 519 | |||

TBL1XR1
Genomic Map