siRNA Details
Gene Symbol: VPS37B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024667.2 | NP_078943.1 | 1, 2 | 219 | ||
GenBank mRNA | AK022812.1 | - | 1, 2 | 219 | ||
BC005882.2 | - | 1, 2 | 157 | 3533384 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: VPS37B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024667.2 | NP_078943.1 | 1, 2 | 219 | ||
GenBank mRNA | AK022812.1 | - | 1, 2 | 219 | ||
BC005882.2 | - | 1, 2 | 157 | 3533384 |