siRNA Details
| Gene Symbol: NSUN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024677.4 | NP_078953.3 | 2 | 668 | ||
| XM_017008611.1 | XP_016864100.1 | 2 | 691 | |||
| XM_017008612.1 | XP_016864101.1 | 2 | 691 | |||
| XM_017008613.1 | XP_016864102.1 | 2 | 691 | |||
| XM_017008614.1 | XP_016864103.1 | 2 | 691 | |||
| XM_017008615.1 | XP_016864104.1 | 2 | 691 | |||
| GenBank mRNA | AK024063.1 | - | 2 | 601 | ||
| AK304048.1 | - | 2 | 392 | |||
| BC036568.2 | - | 2 | 640 | 5272990 | ||
| BC042401.1 | - | 2 | 668 | |||

NSUN7
Genomic Map