siRNA Details
| Gene Symbol: CXorf36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024689.2 | NP_078965.2 | 1 | 238 | ||
| NM_176819.3 | NP_789789.2 | 1 | 238 | |||
| XM_005272670.1 | XP_005272727.1 | 1 | 213 | |||
| XM_006724559.1 | XP_006724622.1 | 1 | 213 | |||
| GenBank mRNA | AK024165.1 | - | 1 | 238 | ||
| AY358764.1 | - | 1 | 196 | |||
| BC035375.1 | - | 1 | 174 | |||
| BC137528.1 | - | 1 | 226 | |||
| BC137529.1 | - | 1 | 226 | |||
| BP231945.1 | - | 1 | 189 | |||

CXorf36
Genomic Map